Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0600005804.mm.1 |
TC0600000775.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600005804.mm.1 |
NM_001177558.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600005804.mm.1 |
NM_001177558.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600005805.mm.1 |
TC0600000775.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600005805.mm.1 |
NM_001177556.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600005805.mm.1 |
NM_001177556.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600005805.mm.1 |
NM_001177557.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600005805.mm.1 |
NM_001177557.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600005805.mm.1 |
NM_001177558.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600005805.mm.1 |
NM_001177558.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600005805.mm.1 |
NM_001177559.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600005805.mm.1 |
NM_001177559.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600005805.mm.1 |
NM_001177560.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600005805.mm.1 |
NM_001177560.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600005805.mm.1 |
NM_025278.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600005805.mm.1 |
NM_025278.5 |
Refseq |