Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gfpt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013528.3 | NP_038556.1 | 1 - 2 | 159 | 97 | |
GenBank mRNA | AF334736.1 | - | 1 - 2 | 116 | 97 | |
AK019852.1 | - | 1 - 2 | 155 | 97 | ||
AK039792.1 | - | 1 - 2 | 157 | 97 | ||
AK087173.1 | - | 1 - 2 | 160 | 97 | ||
BB664274.1 | - | 1 - 2 | 160 | 97 | ||
BC010516.1 | - | 1 - 2 | 107 | 4191553 | 97 | |
BC050762.1 | - | 1 - 2 | 155 | 6742987 | 97 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |