Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Antxr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_054041.2 | NP_473382.1 | 3 - 4 | 624 | 78 | |
GenBank mRNA | AF378762.1 | - | 3 - 4 | 564 | 78 | |
AK031465.1 | - | 3 - 4 | 614 | 78 | ||
AK048637.1 | - | 3 - 4 | 671 | 78 | ||
BC057043.1 | - | 1 - 2 | 62 | 78 | ||
BC094544.1 | - | 3 - 4 | 569 | 6406827 | 78 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |