Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rad52 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001166381.1 | NP_001159853.1 | 9 - 10 | 1042 | 93 | |
NM_001166383.1 | NP_001159855.1 | 9 - 10 | 1042 | 93 | ||
NM_011236.2 | NP_035366.2 | 9 - 10 | 1042 | 93 | ||
GenBank mRNA | BC010673.1 | - | 8 - 9 | 914 | 93 | |
BC019123.1 | - | 9 - 10 | 1022 | 5003696 | 93 | |
U06837.1 | - | 9 - 10 | 964 | 93 | ||
Z32767.1 | - | 9 - 10 | 1036 | 93 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |