Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1000000042.mm.1 |
NM_001033253.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000000042.mm.1 |
NM_001033253.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000000042.mm.1 |
TC1000000011.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000000043.mm.1 |
NM_001033253.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000000043.mm.1 |
NM_001033253.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000000043.mm.1 |
NM_001159942.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000000043.mm.1 |
NM_001159942.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000000043.mm.1 |
TC1000000011.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000000046.mm.1 |
NM_001033253.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000000046.mm.1 |
NM_001033253.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000000046.mm.1 |
NM_001159942.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000000046.mm.1 |
NM_001159942.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000000046.mm.1 |
TC1000000011.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000000041.mm.1 |
NM_001159942.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000000041.mm.1 |
NM_001159942.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000000041.mm.1 |
TC1000000011.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000000045.mm.1 |
NM_001033253.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000000045.mm.1 |
NM_001033253.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000000045.mm.1 |
NM_001159942.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000000045.mm.1 |
NM_001159942.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000000045.mm.1 |
TC1000000011.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000000039.mm.1 |
NM_001159942.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000000039.mm.1 |
NM_001159942.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000000039.mm.1 |
TC1000000011.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000000048.mm.1 |
NM_001033253.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000000048.mm.1 |
NM_001033253.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000000048.mm.1 |
NM_001159942.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000000048.mm.1 |
NM_001159942.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000000048.mm.1 |
TC1000000011.mm.1 |
MTA TC |