Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rpia | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009075.2 | NP_033101.2 | 4 - 5 | 465 | 63 | |
GenBank mRNA | AK137235.1 | - | 4 - 5 | 300 | 63 | |
BC053526.1 | - | 4 - 5 | 288 | 63 | ||
L35034.1 | - | 4 - 5 | 445 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |