Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mfap5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_015776.2 | NP_056591.1 | 8 - 9 | 361 | 87 | |
GenBank mRNA | AF180805.1 | - | 8 - 9 | 361 | 87 | |
AK003479.1 | - | 8 - 9 | 378 | 87 | ||
AK011458.1 | - | 8 - 9 | 438 | 87 | ||
BC025131.1 | - | 8 - 9 | 360 | 3982519 | 87 | |
BC041146.1 | - | 2 - 3 | 2093 | 87 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |