Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Necap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026267.2 | NP_080543.2 | 5 - 6 | 503 | 89 | |
GenBank mRNA | AK004805.1 | - | 5 - 6 | 504 | 89 | |
AK005263.1 | - | 5 - 6 | 494 | 89 | ||
AK081598.1 | - | 5 - 6 | 504 | 89 | ||
AK168500.1 | - | 5 - 6 | 504 | 89 | ||
AK220236.1 | - | 5 - 6 | 489 | 89 | ||
BC011466.1 | - | 5 - 6 | 502 | 4013043 | 89 | |
BC119133.1 | - | 5 - 6 | 515 | 8733882 | 89 | |
BC120841.1 | - | 5 - 6 | 586 | 40129764 | 89 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |