Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Foxj2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021899.3 | NP_068699.1 | 4 - 5 | 1277 | 125 | |
GenBank mRNA | AB221666.1 | - | 3 - 4 | 476 | 125 | |
AF253052.1 | - | 4 - 5 | 1289 | 125 | ||
BC040395.1 | - | 5 - 6 | 907 | 4190087 | 125 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |