Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: A2m | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175628.3 | NP_783327.2 | 22 - 23 | 2892 | 111 | |
GenBank mRNA | AY185125.1 | - | 22 - 23 | 2838 | 111 | |
BC023844.1 | - | 12 - 13 | 1554 | 111 | ||
BC072642.1 | - | 22 - 23 | 2832 | 6839761 | 111 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |