Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Syn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001111015.1 | NP_001104485.1 | 7 - 8 | 1161 | 71 | |
NM_013681.3 | NP_038709.1 | 7 - 8 | 1161 | 71 | ||
GenBank mRNA | AF096867.1 | - | 7 - 8 | 1162 | 71 | |
AK029181.1 | - | 5 - 6 | 503 | 71 | ||
AK043584.1 | - | 2 - 3 | 1373 | 71 | ||
BC053427.1 | - | 7 - 8 | 1082 | 71 | ||
BC066004.1 | - | 7 - 8 | 980 | 71 | ||
BC085129.1 | - | 7 - 8 | 980 | 5701861 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |