Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tsen2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199033.1 | NP_950198.1 | 9 - 10 | 1168 | 65 | |
GenBank mRNA | AK134697.1 | - | 9 - 10 | 1164 | 65 | |
BC061473.1 | - | 9 - 10 | 1168 | 6414790 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |