Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tsen2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199033.1 | NP_950198.1 | 10 - 11 | 1276 | 77 | |
GenBank mRNA | AK134697.1 | - | 10 - 11 | 1272 | 77 | |
BC061473.1 | - | 10 - 11 | 1276 | 6414790 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |