Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Timp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_080639.3 | NP_542370.3 | 1 - 2 | 216 | 133 | |
GenBank mRNA | AK038820.1 | - | 1 - 2 | 215 | 133 | |
AK040814.1 | - | 1 - 2 | 215 | 133 | ||
AK079347.1 | - | 1 - 2 | 217 | 133 | ||
AK140367.1 | - | 1 - 2 | 216 | 133 | ||
BC064046.1 | - | 1 - 2 | 196 | 875422 | 133 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |