Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Foxm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008021.4 | NP_032047.4 | 4 - 5 | 1007 | 83 | |
GenBank mRNA | AK049177.1 | - | 4 - 5 | 948 | 83 | |
AK213171.1 | - | 1 - 2 | 153 | 83 | ||
BC065067.1 | - | 4 - 5 | 1007 | 6417437 | 83 | |
BC075723.1 | - | 4 - 5 | 1026 | 6515008 | 83 | |
Y11245.1 | - | 4 - 5 | 915 | 83 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |