Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0600022404.mm.1 |
TC0600002918.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600022395.mm.1 |
TC0600002918.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600022395.mm.1 |
NM_133932.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600022395.mm.1 |
NM_133932.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600022402.mm.1 |
TC0600002918.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600022402.mm.1 |
NM_133932.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600022402.mm.1 |
NM_133932.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600022400.mm.1 |
TC0600002918.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600022400.mm.1 |
NM_133932.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600022400.mm.1 |
NM_133932.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600022403.mm.1 |
TC0600002918.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600022403.mm.1 |
NM_133932.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600022403.mm.1 |
NM_133932.2 |
Refseq |