Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Camk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133926.2 | NP_598687.1 | 7 - 8 | 755 | 129 | |
GenBank mRNA | AK089742.1 | - | 7 - 8 | 738 | 129 | |
AK134936.1 | - | 7 - 8 | 775 | 129 | ||
AK170188.1 | - | 6 - 7 | 821 | 129 | ||
AK208371.1 | - | 2 - 3 | 237 | 129 | ||
BC014825.1 | - | 7 - 8 | 755 | 3969343 | 129 | |
BC042494.1 | - | 3 - 4 | 2014 | 129 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protein Kinase Mammalian Gene Collection Mus musculus Probe spans exons |