Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Setd5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028385.1 | NP_082661.1 | 15 - 16 | 2540 | 83 | |
GenBank mRNA | AK049143.1 | - | 9 - 10 | 1618 | 83 | |
AK122551.1 | - | 15 - 16 | 2540 | 83 | ||
BC056502.1 | - | 18 - 19 | 2879 | 83 | ||
BC083184.1 | - | 14 - 15 | 2421 | 30536831 | 83 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |