Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ret | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080780.1 | NP_001074249.1 | 8 - 9 | 2268 | 53 | |
NM_009050.2 | NP_033076.2 | 8 - 9 | 2268 | 53 | ||
GenBank mRNA | AF209436.1 | - | 8 - 9 | 1657 | 53 | |
AK044686.1 | - | 8 - 9 | 1855 | 53 | ||
AK051633.1 | - | 8 - 9 | 1847 | 53 | ||
AK084977.1 | - | 8 - 9 | 1918 | 53 | ||
AY326397.1 | - | 8 - 9 | 1657 | 53 | ||
BC059012.1 | - | 8 - 9 | 2242 | 6849696 | 53 | |
X67812.1 | - | 8 - 9 | 1657 | 53 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protein Kinase Mammalian Gene Collection Mus musculus Probe spans exons |