Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0600023138.mm.1 |
TC0600003003.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600023138.mm.1 |
NM_001081112.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600023138.mm.1 |
NM_001081112.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600023136.mm.1 |
TC0600003003.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600023136.mm.1 |
NM_001081112.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600023136.mm.1 |
NM_001081112.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600023139.mm.1 |
TC0600003003.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600023139.mm.1 |
NM_001081112.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600023139.mm.1 |
NM_001081112.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600023137.mm.1 |
TC0600003003.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600023137.mm.1 |
NM_001081112.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600023137.mm.1 |
NM_001081112.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600023140.mm.1 |
TC0600003003.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600023140.mm.1 |
NM_001081112.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600023140.mm.1 |
NM_001081112.1 |
Refseq |