Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bms1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_194339.1 | NP_919320.1 | 13 - 14 | 2505 | 93 | |
GenBank mRNA | AK129082.1 | - | 13 - 14 | 2465 | 93 | |
AK133013.1 | - | 13 - 14 | 2455 | 93 | ||
AK134315.1 | - | 13 - 14 | 2513 | 93 | ||
AK162178.1 | - | 13 - 14 | 2563 | 93 | ||
BC030906.1 | - | 6 - 7 | 1305 | 93 | ||
BC057054.1 | - | 13 - 14 | 2505 | 6827225 | 93 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |