Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aup1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001301649.1 | NP_001288578.1 | 9 - 10 | 1186 | 113 | |
NM_007517.4 | NP_031543.2 | 10 - 11 | 1337 | 113 | ||
GenBank mRNA | AK154707.1 | - | 10 - 11 | 1206 | 113 | |
AK158035.1 | - | 10 - 11 | 1128 | 113 | ||
AK171011.1 | - | 10 - 11 | 1140 | 113 | ||
AK171184.1 | - | 10 - 11 | 1133 | 113 | ||
AK171202.1 | - | 10 - 11 | 1147 | 113 | ||
AK171249.1 | - | 10 - 11 | 1127 | 113 | ||
AK171541.1 | - | 10 - 11 | 1127 | 113 | ||
AK171621.1 | - | 10 - 11 | 1120 | 113 | ||
BC012699.1 | - | 9 - 10 | 970 | 3986517 | 113 | |
BC016485.1 | - | 10 - 11 | 1130 | 3962982 | 113 | |
BM900175.1 | - | 2 - 3 | 113 | |||
U41736.1 | - | 10 - 11 | 1135 | 113 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |