Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aup1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001301649.1 | NP_001288578.1 | 3 - 4 | 637 | 79 | |
NM_007517.4 | NP_031543.2 | 4 - 5 | 788 | 79 | ||
NR_125891.1 | 4 - 5 | 788 | 79 | |||
GenBank mRNA | AK154707.1 | - | 4 - 5 | 657 | 79 | |
AK158035.1 | - | 4 - 5 | 579 | 79 | ||
AK167335.1 | - | 4 - 5 | 578 | 79 | ||
AK171011.1 | - | 4 - 5 | 591 | 79 | ||
AK171184.1 | - | 4 - 5 | 584 | 79 | ||
AK171202.1 | - | 4 - 5 | 598 | 79 | ||
AK171249.1 | - | 4 - 5 | 578 | 79 | ||
AK171541.1 | - | 4 - 5 | 578 | 79 | ||
AK171621.1 | - | 4 - 5 | 571 | 80 | ||
BC012699.1 | - | 3 - 4 | 421 | 3986517 | 79 | |
BC016485.1 | - | 4 - 5 | 581 | 3962982 | 79 | |
U41736.1 | - | 4 - 5 | 586 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |