Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aup1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001301649.1 | NP_001288578.1 | 8 - 9 | 1069 | 71 | |
NM_007517.4 | NP_031543.2 | 9 - 10 | 1220 | 71 | ||
NR_125891.1 | 9 - 10 | 1220 | 71 | |||
GenBank mRNA | AK154707.1 | - | 9 - 10 | 1089 | 71 | |
AK158035.1 | - | 9 - 10 | 1011 | 71 | ||
AK171011.1 | - | 9 - 10 | 1023 | 71 | ||
AK171184.1 | - | 9 - 10 | 1016 | 71 | ||
AK171202.1 | - | 9 - 10 | 1030 | 71 | ||
AK171249.1 | - | 9 - 10 | 1010 | 71 | ||
AK171541.1 | - | 9 - 10 | 1010 | 71 | ||
AK171621.1 | - | 9 - 10 | 1003 | 71 | ||
AK188454.1 | - | 3 - 4 | 258 | 71 | ||
BC012699.1 | - | 8 - 9 | 853 | 3986517 | 71 | |
BC016485.1 | - | 9 - 10 | 1013 | 3962982 | 71 | |
U41736.1 | - | 9 - 10 | 1018 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |