Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tet3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_183138.2 | NP_898961.2 | 1 - 2 | 131 | 60 | |
GenBank mRNA | AK044758.1 | - | 1 - 2 | 84 | 60 | |
AK046543.1 | - | 1 - 2 | 132 | 60 | ||
AK046552.1 | - | 1 - 2 | 132 | 60 | ||
AK046553.1 | - | 1 - 2 | 132 | 60 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Detect Genomic DNA Mus musculus Probe spans exons |