Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pex3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164195.1 | NP_001157667.1 | 7 - 8 | 822 | 146 | |
NM_019961.3 | NP_064345.1 | 7 - 8 | 822 | 146 | ||
GenBank mRNA | AF152996.1 | - | 7 - 8 | 745 | 146 | |
AK005315.1 | - | 7 - 8 | 678 | 146 | ||
AK005978.1 | - | 6 - 7 | 519 | 146 | ||
AK012827.1 | - | 7 - 8 | 801 | 146 | ||
AK075820.1 | - | 7 - 8 | 737 | 146 | ||
AK083778.1 | - | 2 - 3 | 309 | 146 | ||
AK146934.1 | - | 7 - 8 | 801 | 146 | ||
BC033415.1 | - | 7 - 8 | 704 | 5008068 | 146 | |
BC037606.1 | - | 7 - 8 | 785 | 5361347 | 146 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |