Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ltv1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_181470.4 | NP_852135.1 | 7 - 8 | 1010 | 79 | |
GenBank mRNA | AK011491.1 | - | 7 - 8 | 996 | 79 | |
BC067062.1 | - | 7 - 8 | 1010 | 30538957 | 79 | |
BC069962.1 | - | 7 - 8 | 1004 | 5038459 | 79 | |
U01139.1 | - | 4 - 5 | 585 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |