Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptpn6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077705.2 | NP_001071173.1 | 8 - 9 | 1133 | 77 | |
NM_013545.3 | NP_038573.2 | 8 - 9 | 1131 | 77 | ||
GenBank mRNA | AK132509.1 | - | 8 - 9 | 1050 | 77 | |
AK150509.1 | - | 8 - 9 | 1038 | 77 | ||
AK151077.1 | - | 8 - 9 | 1052 | 77 | ||
AK151623.1 | - | 8 - 9 | 1055 | 77 | ||
AK159686.1 | - | 8 - 9 | 1090 | 77 | ||
AK159833.1 | - | 8 - 9 | 1083 | 77 | ||
AK167923.1 | - | 8 - 9 | 1085 | 77 | ||
AK169804.1 | - | 8 - 9 | 1133 | 77 | ||
BC012660.1 | - | 8 - 9 | 1076 | 4010462 | 77 | |
M68902.1 | - | 8 - 9 | 1083 | 77 | ||
M90389.1 | - | 8 - 9 | 1033 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protein Phosphatase Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |