Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cops7a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012003.2 | NP_036133.1 | 7 - 8 | 947 | 68 | |
GenBank mRNA | AF071316.1 | - | 7 - 8 | 949 | 68 | |
BC003724.1 | - | 6 - 7 | 828 | 3593979 | 68 | |
CF898433.1 | - | 3 - 4 | 443 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |