Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pex5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277330.1 | NP_001264259.1 | 5 - 6 | 560 | 94 | |
NM_001277805.1 | NP_001264734.1 | 5 - 6 | 558 | 94 | ||
NM_008995.2 | NP_033021.2 | 5 - 6 | 521 | 94 | ||
NM_175933.2 | NP_787947.1 | 5 - 6 | 558 | 94 | ||
GenBank mRNA | AJ416473.1 | - | 5 - 6 | 540 | 94 | |
AK088886.1 | - | 5 - 6 | 562 | 94 | ||
AK145111.1 | - | 5 - 6 | 494 | 94 | ||
AK145361.1 | - | 5 - 6 | 495 | 94 | ||
AK161470.1 | - | 5 - 6 | 555 | 94 | ||
BC029748.1 | - | 5 - 6 | 528 | 2599159 | 94 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |