Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clstn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153508.4 | NP_705728.1 | 8 - 9 | 1622 | 67 | |
GenBank mRNA | AJ278486.1 | - | 8 - 9 | 1489 | 67 | |
AK032336.1 | - | 8 - 9 | 1564 | 67 | ||
BC041781.1 | - | 8 - 9 | 1321 | 67 | ||
BC055054.1 | - | 8 - 9 | 1554 | 6493096 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |