Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcnq4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081142.1 | NP_001074611.1 | 5 - 6 | 840 | 82 | |
GenBank mRNA | AK040190.1 | - | 5 - 6 | 773 | 82 | |
AK163659.1 | - | 629 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Mus musculus Probe spans exons |