Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stil | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009185.3 | NP_033211.2 | 4 - 5 | 542 | 88 | |
GenBank mRNA | AK049223.1 | - | 4 - 5 | 479 | 88 | |
AK049550.1 | - | 4 - 5 | 479 | 88 | ||
BC049865.1 | - | 4 - 5 | 496 | 88 | ||
BC060706.1 | - | 4 - 5 | 457 | 88 | ||
BC087545.1 | - | 4 - 5 | 481 | 88 | ||
BC141175.1 | - | 4 - 5 | 415 | 9055787 | 88 | |
BC145493.1 | - | 4 - 5 | 415 | 9054056 | 88 | |
U36778.1 | - | 4 - 5 | 460 | 88 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |