Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eif2b3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001111277.1 | NP_001104747.1 | 3 - 4 | 376 | 140 | |
NM_175135.4 | NP_780344.2 | 3 - 4 | 376 | 140 | ||
GenBank mRNA | AK145856.1 | - | 3 - 4 | 353 | 140 | |
AK172173.1 | - | 3 - 4 | 370 | 140 | ||
AK172302.1 | - | 3 - 4 | 326 | 140 |
Set Membership: |
Amplicon length greater than or equal to 101 Mus musculus Probe spans exons |