Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atp6ap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_018794.4 | NP_061264.1 | 1 - 2 | 225 | 93 | |
GenBank mRNA | AB031290.1 | - | 1 - 2 | 143 | 93 | |
AK093961.1 | - | 1 - 2 | 195 | 93 | ||
AK150333.1 | - | 1 - 2 | 160 | 93 | ||
AK158956.1 | - | 1 - 2 | 213 | 93 | ||
AK159449.1 | - | 1 - 2 | 176 | 93 | ||
AK159612.1 | - | 1 - 2 | 157 | 93 | ||
AK161631.1 | - | 1 - 2 | 164 | 93 | ||
AK166793.1 | - | 1 - 2 | 159 | 93 | ||
AK220247.1 | - | 1 - 2 | 216 | 93 | ||
BC048241.1 | - | 1 - 2 | 177 | 6493219 | 93 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Transporter Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons |