Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gatsl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028022.1 | NP_082298.1 | 6 - 7 | 803 | 89 | |
GenBank mRNA | AK010455.1 | - | 6 - 7 | 786 | 89 | |
BC039988.1 | - | 6 - 7 | 803 | 1195616 | 89 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |