Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gatsl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK010455.1 | - | 7 - 8 | 870 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Mus musculus Probe spans exons |