Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tbc1d10a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_134023.1 | NP_598784.1 | 3 - 4 | 473 | 64 | |
GenBank mRNA | AK140872.1 | - | 3 - 4 | 485 | 64 | |
AK153677.1 | - | 455 | 64 | |||
AK197604.1 | - | 1 - 2 | 65 | 64 | ||
AK220243.1 | - | 484 | 64 | |||
BC018300.1 | - | 3 - 4 | 473 | 4193046 | 64 | |
BC029773.1 | - | 1 - 2 | 37 | 64 | ||
BC037230.1 | - | 3 - 4 | 456 | 2650923 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |