Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pik3ip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK190351.1 | - | 1 - 2 | 75 | 57 |
Set Membership: |
Amplicon length less than or equal to 70 Mus musculus Probe spans exons |