Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zpbp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001185153.1 | NP_001172082.1 | 2 - 3 | 268 | 109 | |
NM_015785.2 | NP_056600.1 | 2 - 3 | 439 | 109 | ||
GenBank mRNA | BC139450.1 | - | 2 - 3 | 408 | 8862472 | 109 |
BC139453.1 | - | 2 - 3 | 408 | 8862475 | 109 | |
BC145628.1 | - | 2 - 3 | 237 | 9054196 | 109 | |
D17569.1 | - | 2 - 3 | 428 | 109 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |