Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zpbp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001185153.1 | NP_001172082.1 | 6 - 7 | 838 | 86 | |
NM_015785.2 | NP_056600.1 | 6 - 7 | 1009 | 86 | ||
GenBank mRNA | AK006274.1 | - | 3 - 4 | 351 | 86 | |
AK018889.1 | - | 3 - 4 | 353 | 86 | ||
AK018921.1 | - | 3 - 4 | 353 | 86 | ||
AK029541.1 | - | 3 - 4 | 353 | 86 | ||
BC139450.1 | - | 6 - 7 | 978 | 8862472 | 86 | |
BC139453.1 | - | 6 - 7 | 978 | 8862475 | 86 | |
BC145628.1 | - | 6 - 7 | 807 | 9054196 | 86 | |
D17569.1 | - | 6 - 7 | 998 | 86 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |