Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cobl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001282993.1 | NP_001269922.1 | 4 - 5 | 928 | 67 | |
NM_001282994.1 | NP_001269923.1 | 4 - 5 | 759 | 67 | ||
NM_172496.3 | NP_766084.3 | 4 - 5 | 928 | 67 | ||
GenBank mRNA | AK028833.1 | - | 4 - 5 | 775 | 67 | |
AK049730.1 | - | 4 - 5 | 721 | 67 | ||
AK122334.1 | - | 4 - 5 | 759 | 67 | ||
AK147366.1 | - | 4 - 5 | 928 | 67 | ||
AK147633.1 | - | 4 - 5 | 928 | 67 | ||
AY308745.1 | - | 4 - 5 | 768 | 67 | ||
BC145116.1 | - | 4 - 5 | 768 | 67 | ||
BC145118.1 | - | 4 - 5 | 768 | 9053666 | 67 | |
BC150706.1 | - | 4 - 5 | 768 | 9087617 | 67 | |
U26967.1 | - | 3 - 4 | 501 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |