Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0400026872.mm.1 |
TC0400003360.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400026872.mm.1 |
NR_024078.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400026872.mm.1 |
NR_024078.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400026873.mm.1 |
TC0400003360.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400026873.mm.1 |
NR_024078.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400026873.mm.1 |
NR_024078.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400026876.mm.1 |
TC0400003360.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400026876.mm.1 |
NR_024078.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400026876.mm.1 |
NR_024078.1 |
Refseq |