Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tcf7l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079822.2 | NP_001073290.1 | 7 - 8 | 889 | 83 | |
NM_009332.3 | NP_033358.2 | 6 - 7 | 847 | 83 | ||
GenBank mRNA | AF057691.1 | - | 2 - 3 | 235 | 83 | |
AJ223069.1 | - | 6 - 7 | 755 | 83 | ||
AJ223233.1 | - | 6 - 7 | 670 | 83 | ||
AK052721.1 | - | 6 - 7 | 1057 | 83 | ||
BC128305.1 | - | 7 - 8 | 796 | 40109611 | 83 | |
BC128306.1 | - | 6 - 7 | 754 | 40109613 | 83 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |