Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0400008892.mm.1 |
NM_001100183.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400008892.mm.1 |
NM_001100183.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400008892.mm.1 |
TC0400001220.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400008893.mm.1 |
NM_001100183.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400008893.mm.1 |
NM_001100183.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400008893.mm.1 |
TC0400001220.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400008888.mm.1 |
NM_001100183.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400008888.mm.1 |
NM_001100183.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400008888.mm.1 |
TC0400001220.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400008899.mm.1 |
NM_001100183.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400008899.mm.1 |
NM_001100183.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400008899.mm.1 |
TC0400001220.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400008895.mm.1 |
NM_001100183.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400008895.mm.1 |
NM_001100183.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400008895.mm.1 |
TC0400001220.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400008898.mm.1 |
NM_001100183.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400008898.mm.1 |
NM_001100183.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400008898.mm.1 |
TC0400001220.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400008889.mm.1 |
NM_001100183.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400008889.mm.1 |
NM_001100183.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400008889.mm.1 |
TC0400001220.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400008896.mm.1 |
NM_001100183.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400008896.mm.1 |
NM_001100183.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400008896.mm.1 |
TC0400001220.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400008891.mm.1 |
NM_001100183.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400008891.mm.1 |
NM_001100183.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400008891.mm.1 |
TC0400001220.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400008894.mm.1 |
NM_001100183.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400008894.mm.1 |
NM_001100183.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400008894.mm.1 |
TC0400001220.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400008897.mm.1 |
NM_001100183.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400008897.mm.1 |
NM_001100183.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400008897.mm.1 |
TC0400001220.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400008890.mm.1 |
NM_001100183.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400008890.mm.1 |
NM_001100183.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400008890.mm.1 |
TC0400001220.mm.1 |
MTA TC |