Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cand2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025958.2 | NP_080234.2 | 13 - 14 | 3427 | 58 | |
GenBank mRNA | AK129186.1 | - | 13 - 14 | 3398 | 58 | |
AK145112.1 | - | 13 - 14 | 3412 | 58 | ||
AK170420.1 | - | 4 - 5 | 1131 | 58 | ||
BC058338.1 | - | 12 - 13 | 3242 | 58 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |