Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cand2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025958.2 | NP_080234.2 | 7 - 8 | 1057 | 67 | |
GenBank mRNA | AK129186.1 | - | 7 - 8 | 1028 | 67 | |
BC056365.1 | - | 7 - 8 | 1039 | 6824847 | 67 | |
BC058338.1 | - | 6 - 7 | 872 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |