Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ctnna2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109764.1 | NP_001103234.1 | 9 - 10 | 1492 | 123 | |
NM_009819.2 | NP_033949.2 | 8 - 9 | 1390 | 123 | ||
NM_145732.2 | NP_663785.2 | 9 - 10 | 1492 | 123 | ||
GenBank mRNA | AK158916.1 | - | 9 - 10 | 1525 | 123 | |
BC051995.1 | - | 8 - 9 | 1271 | 123 | ||
BC079648.1 | - | 9 - 10 | 1409 | 6850068 | 123 | |
D25281.1 | - | 9 - 10 | 1419 | 123 | ||
D25282.1 | - | 8 - 9 | 1319 | 123 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |