Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Alms1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145223.2 | NP_660258.2 | 11 - 12 | 7280 | 150 | |
GenBank mRNA | AF425257.1 | - | 11 - 12 | 7281 | 150 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mus musculus Probe spans exons |