Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdc40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027879.2 | NP_082155.1 | 4 - 5 | 557 | 144 | |
GenBank mRNA | AK004569.1 | - | 4 - 5 | 524 | 144 | |
AK040680.1 | - | 4 - 5 | 507 | 144 | ||
AK135411.1 | - | 4 - 5 | 524 | 144 | ||
BC061220.1 | - | 4 - 5 | 532 | 144 | ||
BC147503.1 | - | 4 - 5 | 557 | 9056386 | 144 | |
BC158078.1 | - | 4 - 5 | 557 | 9088170 | 144 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |